3-(2-Pyridyl)benzaldehyde ≥97.0% (by GC)

Synonyms: 3-(Pyridin-2-yl)benzaldehyde

P1889-1G
TCP1889-1GEA 356.31 USD
TCP1889-1G
3-(2-Pyridyl)benzaldehyde ≥97.0% (by GC)
3-(2-Pyridyl)benzaldehyde

CAS Number: 85553-53-3
MDL Number: MFCD02684103
Molecular Formula: C12H9NO
Molecular Weight: 183.21
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Color: Slightly Pale Yellow


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors, Ligands for Functional Metal Complexes
Other Product Names / Synonyms: 2-(3-Formylphenyl)pyridine

Formula: C₁₂H₉NO
MW: 183.2060 g/mol
MDL Number: MFCD02684103
CAS Number: 85553-53-3
UN: 2810
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/31/67/28993167.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR