Prostaglandin E2 ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
PGE2
P1884-10MG
P1884-1MG
TCP1884-10MGEA
450.64
USD
TCP1884-10MG
TCP1884-1MG
Prostaglandin E2 ≥98.0% (by HPLC)
Prostaglandin E2
CAS Number: 363-24-6
MDL Number: MFCD00077861
Molecular Formula: C20H32O5
Molecular Weight: 352.47
Purity/Analysis Method:
Form: Crystal
Color: White
Melting point (°C): 66
Storage Temperature:
Product Categories: Bioscience, Chemical Mediators, Reagents for Pain & Itch Models, Lipid Mediators, Eicosanoids, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: PGE2
Formula:
C₂₀H₃₂O₅ MW: 352.47 g/mol Melting Pt: 66 °C |
MDL Number:
MFCD00077861 CAS Number: 363-24-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...