Propyne
Supplier: TCI America
Synonyms:
Methylacetylene
P1881-200G
TCP1881-200GEA
211.12
USD
TCP1881-200G
Propyne
Propyne
CAS Number: 74-99-7
MDL Number: MFCD00036235
Molecular Formula: C3H4
Molecular Weight: 40.07
Form: Clear Liquid
Color: Colorless
Storage Temperature:
Other Product Names / Synonyms: Methylacetylene
Formula:
C₃H₄ MW: 40.06 g/mol |
MDL Number:
MFCD00036235 CAS Number: 74-99-7 UN: 1954 ADR: 2, |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...