6-Phenoxyhexyl bromide ≥96.0% (by GC)
Supplier: TCI America
Synonyms:
6-Bromohexyl Phenyl Ether
P1848-5G
TCP1848-5GEA
163
USD
TCP1848-5G
6-Phenoxyhexyl bromide ≥96.0% (by GC)
6-Phenoxyhexyl bromide
CAS Number: 51795-97-2
MDL Number: MFCD00039205
Molecular Formula: C12H17BrO
Molecular Weight: 257.17
Purity/Analysis Method:
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 112
Specific Gravity (20/20): 1.25
Other Product Names / Synonyms: 6-Bromohexyl Phenyl Ether
Formula:
C₁₂H₁₇BrO MW: 257.17 g/mol Boiling Pt: 112 °C Density: 1.25 (20 °C) |
MDL Number:
MFCD00039205 CAS Number: 51795-97-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...