5-Phenoxyamyl bromide ≥95.0% (by GC)
Supplier: TCI America
Synonyms:
5-Bromopentyl Phenyl Ether
, 5-Phenoxypentyl Bromide
, 5-Bromoamyl Phenyl Ether
CAS Number: 22921-72-8
MDL Number: MFCD00039203
Molecular Formula: C11H15BrO
Molecular Weight: 243.14
Purity/Analysis Method:
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 130
Specific Gravity (20/20): 1.28
Other Product Names / Synonyms: 5-Phenoxypentyl Bromide, 5-Bromoamyl Phenyl Ether, 5-Bromopentyl Phenyl Ether
Formula:
C₁₁H₁₅BrO MW: 243.14 g/mol Boiling Pt: 130 °C Density: 1.28 (20 °C) |
MDL Number:
MFCD00039203 CAS Number: 22921-72-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...