(Pentamethylcyclopentadienyl)titanium trichloride ≥97.0% (by titration, gravimetric analysis)
Supplier: TCI America
Synonyms:
Trichloro(pentamethylcyclopentadienyl)titanium(IV)
P1651-1G
TCP1651-1GEA
83.2
USD
TCP1651-1G
(Pentamethylcyclopentadienyl)titanium trichloride ≥97.0% (by titration, gravimetric analysis)
(Pentamethylcyclopentadienyl)titanium trichloride
CAS Number: 12129-06-5
Molecular Formula: C10H15Cl3Ti
Molecular Weight: 289.45
Purity/Analysis Method:
Form: Crystal
Color: Deep Yellow Red
Melting point (°C): 200
Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Homogeneous Catalysts, Materials Chemistry, Polymerization Catalysts
Other Product Names / Synonyms: Trichloro(pentamethylcyclopentadienyl)titanium(IV)
Formula:
C₁₀H₁₅Cl₃Ti MW: 289.45 g/mol Melting Pt: 200 °C |
MDL Number:
MFCD00070434 CAS Number: 12129-06-5 UN: 3261 ADR: 8,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...