Phenylmagnesium chloride 27% ∼2 mol/L in THF
P1381-250G
TCP1381-250GEA
169.58
USD
TCP1381-250G
Phenylmagnesium chloride 27% ∼2 mol/L in THF
Phenylmagnesium chloride
CAS Number: 100-59-4
MDL Number: MFCD00000464
Molecular Formula: C6H5ClMg
Molecular Weight: 136.86
Flash Point (°C): -17
Specific Gravity (20/20): 1.04
Formula:
C₆H₅ClMg Density: 1,04 (20 °C) Flash Pt: –17 °C |
MDL Number:
MFCD00000464 CAS Number: 100-59-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...