Benzeneselenenyl bromide ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Bromoselenobenzene Phenylselenenyl bromide

P1322-5G P1322-25G
TCP1322-5GEA 149 USD
TCP1322-5G TCP1322-25G
Benzeneselenenyl bromide ≥97.0% (by titrimetric analysis)
Benzeneselenenyl bromide

CAS Number: 34837-55-3
MDL Number: MFCD00000047
Molecular Formula: C6H5BrSe
Molecular Weight: 235.97
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Color: Yellow Red
Boiling point (°C): 107
Melting point (°C): 58


Product Categories: Synthetic Organic Chemistry, Selenenylation
Other Product Names / Synonyms: Benzeneselenenyl Bromide

Formula: (C₆H₅)SeBr
MW: 235.97 g/mol
Boiling Pt: 107 °C
Melting Pt: 58 °C
MDL Number: MFCD00000047
CAS Number: 34837-55-3
UN: 2928
ADR: 6.1,II
https://us.vwr.com/stibo/web/std.lang.all/19/80/28991980.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR