O-Phenyl chlorothioformate ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
Phenyl chlorothionoformate
, Phenyl chlorothionocarbonate
, O-Phenyl carbonochloridothioate
CAS Number: 1005-56-7
MDL Number: MFCD00004920
Molecular Formula: C7H5ClOS
Molecular Weight: 172.63
Purity/Analysis Method:
Form: Clear Liquid
Color: Pale Yellow
Boiling point (°C): 93
Flash Point (°C): 81
Specific Gravity (20/20): 1.28
Product Categories: Synthetic Organic Chemistry, Thiocarbonyl Compounds
Other Product Names / Synonyms: Chlorothioformic Acid O-Phenyl Ester, Phenyl Thionochloroformate
Formula:
C₇H₅ClOS MW: 172.63 g/mol Boiling Pt: 93 °C Density: 1.28 (20 °C) Flash Pt: 81 °C |
MDL Number:
MFCD00004920 CAS Number: 1005-56-7 UN: 3265 ADR: 8,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...