1,3-Propanedisulfonic acid 50 - 60% in water
Supplier: TCI America
P1204-10G
TCP1204-10GEA
79.14
USD
TCP1204-10G
1,3-Propanedisulfonic acid 50 - 60% in water
1,3-Propanedisulfonic acid
CAS Number: 21668-77-9
MDL Number: MFCD00191482
Molecular Formula: C3H8O6S2
Molecular Weight: 204.21
Form: Clear Liquid
Color: Colorless
Formula:
C₃H₈O₆S₂ MW: 204.21 |
MDL Number:
MFCD00191482 CAS Number: 21668-77-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...