O-Phenylenediamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 1,2-PhenylendiamineOPD1,2-Diaminobenzene1,2-Phenylenediamine

P0968-25G
TCP0968-25GEA 132.86 USD
TCP0968-25G
O-Phenylenediamine ≥98.0% (by GC, titration analysis)
o-Phenylenediamine

CAS Number: 95-54-5
MDL Number: MFCD00007721
Molecular Formula: C6H8N2
Molecular Weight: 108.14
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 265
Melting point (°C): 103
Flash Point (°C): 156
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 1,2-Diaminobenzene

Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 265 °C
Melting Pt: 103 °C
Flash Pt: 156 °C
MDL Number: MFCD00007721
CAS Number: 95-54-5
UN: 1673
ADR: 6.1,III

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR