N,N-Dimethylaminophosphoryl dichloride ≥85.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: N,N-Dimethylphosphoramidodichloridate

P0953-25G P0953-5G
TCP0953-5GEA 89.32 USD
TCP0953-5G TCP0953-25G
N,N-Dimethylaminophosphoryl dichloride ≥85.0% (by titrimetric analysis)
N,N-Dimethylaminophosphoryl dichloride

CAS Number: 677-43-0
MDL Number: MFCD00013896
Molecular Formula: C2H6Cl2NOP
Molecular Weight: 161.95
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 195
Flash Point (°C): 110
Specific Gravity (20/20): 1.37


Other Product Names / Synonyms: N,N-Dimethylphosphoramidic Dichloride, Phosphoric Acid Dimethylamide Dichloride

Formula: C₂H₆Cl₂NOP
Boiling Pt: 195 °C
Density: 1.37 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00013896
CAS Number: 677-43-0
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR