(S)-(-)-1,2-Epoxypropane ≥98.0% (by GC)

Supplier: TCI America

Synonyms: (S)-(-)-Methyloxirane (S)-(-)-Propene oxide (S)-(-)-Propylene oxide

P0951-1ML P0951-5ML P0951-25ML
TCP0951-25MLEA 184.68 USD
TCP0951-25ML TCP0951-1ML TCP0951-5ML
(S)-(-)-1,2-Epoxypropane ≥98.0% (by GC)
(S)-(-)-1,2-Epoxypropane

CAS Number: 16088-62-3
MDL Number: MFCD00064312
Molecular Formula: C3H6O
Molecular Weight: 58.08
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 34
Flash Point (°C): -37
Specific Gravity (20/20): 0.83
Specific rotation [a]20/D: -14 deg (neat)
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-(-)-1,2-Epoxypropane

Formula: C₃H₆O
MW: 58.08 g/mol
Boiling Pt: 34 °C
Density: 0.83 (20 °C)
Flash Pt: –37 °C
MDL Number: MFCD00064312
CAS Number: 16088-62-3
UN: 1280
ADR: 3,I
https://us.vwr.com/stibo/web/std.lang.all/16/11/28991611.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR