Pentafluorobenzonitrile ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2,3,4,5,6-Pentafluorobenzonitrile

P0935-25G
TCP0935-25GEA 53.47 USD
TCP0935-25G
Pentafluorobenzonitrile ≥98.0% (by GC)
Pentafluorobenzonitrile

CAS Number: 773-82-0
MDL Number: MFCD00001775
Molecular Formula: C7F5N
Molecular Weight: 193.08
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 162
Flash Point (°C): 29
Specific Gravity (20/20): 1.57


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Building Blocks ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: Perfluorobenzonitrile

Formula: C₇F₅N
MW: 193.08 g/mol
Boiling Pt: 162 °C
Density: 1.57 (20 °C)
Flash Pt: 29 °C
MDL Number: MFCD00001775
CAS Number: 773-82-0
UN: 1992
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/15/98/28991598.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR