Pentafluoroanisole ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2,3,4,5,6-Pentafluoroanisole

P0918-25G P0918-5G
TCP0918-25GEA 280.14 USD
TCP0918-25G TCP0918-5G
Pentafluoroanisole ≥98.0% (by GC)
Pentafluoroanisole

CAS Number: 389-40-2
MDL Number: MFCD00000289
Molecular Formula: C7H3F5O
Molecular Weight: 198.09
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 138
Flash Point (°C): 32
Specific Gravity (20/20): 1.50


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Methoxypentafluorobenzene

Formula: C₇H₃F₅O
MW: 198.09 g/mol
Boiling Pt: 138 °C
Density: 1.50 (20 °C)
Flash Pt: 32 °C
MDL Number: MFCD00000289
CAS Number: 389-40-2
UN: 1993
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/15/81/28991581.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR