1,10-Phenanthroline monohydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 1,10-Phenanthroline hydrate

P0879-25G
TCP0879-25GEA 98.25 USD
TCP0879-25G
1,10-Phenanthroline monohydrate ≥98.0% (by titrimetric analysis)
1,10-Phenanthroline monohydrate

CAS Number: 5144-89-8
MDL Number: MFCD00149973
Molecular Formula: C12H8N2
Molecular Weight: 180.21
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 102

Formula: C₁₂H₈N₂·1H₂O
MW: 198.22 g/mol
Melting Pt: 102 °C
MDL Number: MFCD00011678
CAS Number: 5144-89-8
UN: 2811
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/15/48/28991548.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR