Perfluoro(methylcyclohexane) ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
FlutecR PP2
P0846-100G
P0846-25G
TCP0846-100GEA
267
USD
TCP0846-100G
TCP0846-25G
Perfluoro(methylcyclohexane) ≥98.0% (by GC)
Perfluoro(methylcyclohexane)
CAS Number: 355-02-2
MDL Number: MFCD00001468
Molecular Formula: C7F14
Molecular Weight: 350.05
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 76
Specific Gravity (20/20): 1.80
Product Categories: Synthetic Organic Chemistry, Fluorous Chemistry, Fluorous Compounds
Other Product Names / Synonyms: Perfluoromethylcyclohexane
Formula:
C₇F₁₄ MW: 350.05 g/mol Boiling Pt: 76 °C Density: 1.80 (20 °C) |
MDL Number:
MFCD00001468 CAS Number: 355-02-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...