trans-1,3-Pentadiene ≥95.0% (by GC) stabilized

Supplier: TCI America

Synonyms: trans-Piperylene

P0647-5ML P0647-1ML
TCP0647-5MLEA 264.37 USD
TCP0647-5ML TCP0647-1ML
trans-1,3-Pentadiene ≥95.0% (by GC) stabilized
trans-1,3-Pentadiene

(stabilized with TBC)
CAS Number: 2004-70-8
MDL Number: MFCD00009295
Molecular Formula: C5H8
Molecular Weight: 68.12
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 42
Flash Point (°C): -29
Specific Gravity (20/20): 0.68
Storage Temperature: 0-10°C


Other Product Names / Synonyms: trans-Piperylene

https://us.vwr.com/stibo/web/std.lang.all/13/67/28991367.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR