3-Phenylpropionitrile ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
Hydrocinnamonitrile
P0611-25G
TCP0611-25GEA
77.11
USD
TCP0611-25G
3-Phenylpropionitrile ≥98.0% (by GC)
3-Phenylpropionitrile
CAS Number: 645-59-0
MDL Number: MFCD00001961
Molecular Formula: C9H9N
Molecular Weight: 131.18
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 254
Flash Point (°C): 113
Specific Gravity (20/20): 1.00
Product Categories: Synthetic Organic Chemistry, Building Blocks ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: Hydrocinnamonitrile
Formula:
C₉H₉N MW: 131.18 g/mol Boiling Pt: 254 °C Density: 1.00 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00001961 CAS Number: 645-59-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...