Allylbenzene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 3-Phenylpropene1-Phenyl-2-propene

P0609-25ML P0609-100ML
TCP0609-25MLEA 58.3 USD
TCP0609-25ML TCP0609-100ML
Allylbenzene ≥98.0% (by GC)
Allylbenzene

CAS Number: 300-57-2
MDL Number: MFCD00008651
Molecular Formula: C9H10
Molecular Weight: 118.18
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 155
Flash Point (°C): 33
Specific Gravity (20/20): 0.89


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 3-Phenyl-1-propene

Formula: (C₆H₅)CH₂CHCH₂
Boiling Pt: 155 °C
Density: 0.89 (20 °C)
Flash Pt: 33 °C
MDL Number: MFCD00008651
CAS Number: 300-57-2
UN: 3295
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/13/39/28991339.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR