Primuline
Supplier: TCI America
P0603-5G
TCP0603-5GEA
283.19
USD
TCP0603-5G
Primuline
Primuline
CAS Number: 8064-60-6
MDL Number: MFCD00005798
Form: Crystal
Color: Yellow
MDL Number:
MFCD00005798 CAS Number: 8064-60-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...