3-Pyrroline ≥70.0% (by GC) (contains Pyrrolidine)
Supplier: TCI America
Synonyms:
2,5-Dihydro-1H-pyrrole
P0577-5ML
TCP0577-5MLEA
196
USD
TCP0577-5ML
3-Pyrroline ≥70.0% (by GC) (contains Pyrrolidine)
3-Pyrroline
CAS Number: 109-96-6
MDL Number: MFCD00005213
Molecular Formula: C4H7N
Molecular Weight: 69.11
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 91
Flash Point (°C): -18
Specific Gravity (20/20): 0.91
Formula:
C₄H₇N MW: 69.11 g/mol Boiling Pt: 91 °C Density: 0.91 (20 °C) Flash Pt: –18 °C |
MDL Number:
MFCD00005213 CAS Number: 109-96-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...