Dipropyl sulfide ≥98.0% (by GC)

Supplier: TCI America

P0532-25ML
TCP0532-25MLEA 53.8 USD
TCP0532-25ML
Dipropyl sulfide ≥98.0% (by GC)
Dipropyl sulphide

CAS Number: 111-47-7
MDL Number: MFCD00009379
Molecular Formula: C6H14S
Molecular Weight: 118.24
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 142
Melting point (°C): 0
Flash Point (°C): 31
Specific Gravity (20/20): 0.84


Other Product Names / Synonyms: Dipropyl Sulfide

Formula: C₆H₁₄S
MW: 118.24 g/mol
Boiling Pt: 142 °C
Density: 0.84 (20 °C)
Flash Pt: 31 °C
MDL Number: MFCD00009379
CAS Number: 111-47-7
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR