1-Pentadecyne ≥95.0% (by GC)

Supplier: TCI America

P0356-5ML
TCP0356-5MLEA 175.19 USD
TCP0356-5ML
1-Pentadecyne ≥95.0% (by GC)
1-Pentadecyne

CAS Number: 765-13-9
MDL Number: MFCD00015081
Molecular Formula: C15H28
Molecular Weight: 208.39
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 268
Flash Point (°C): 90
Specific Gravity (20/20): 0.79


Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: Tridecylacetylene

Formula: C₁₅H₂₈
MW: 208.39 g/mol
Boiling Pt: 268 °C
Density: 0.79 (20 °C)
Flash Pt: 90 °C
MDL Number: MFCD00015081
CAS Number: 765-13-9
https://us.vwr.com/stibo/web/std.lang.all/10/83/28991083.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR