Pyrocatechol ≥99.0% (by GC), Technical Grade
Supplier: TCI America
Synonyms:
o-Dihydroxybenzene
CAS Number: 120-80-9
MDL Number: MFCD00002188
Molecular Formula: C6H6O2
Molecular Weight: 110.11
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 246
Melting point (°C): 106
Flash Point (°C): 127
Product Categories: Materials Chemistry, Ligands for Functional Metal Complexes
Other Product Names / Synonyms: 1,2-Dihydroxybenzene, Pyrocatechol
Formula:
C₆H₆O₂ MW: 110.11 g/mol Boiling Pt: 246 °C Melting Pt: 106 °C Flash Pt: 127 °C |
MDL Number:
MFCD00002188 CAS Number: 120-80-9 UN: 2811 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...