1,5-Pentamethylene-1H-tetrazole ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Pentylenetetrazole

P0046-25G
TCP0046-25GEA 156 USD
TCP0046-25G
1,5-Pentamethylene-1H-tetrazole ≥98.0% (by GC)
1,5-Pentamethylene-1H-tetrazole

CAS Number: 54-95-5
MDL Number: MFCD00005939
Molecular Formula: C6H10N4
Molecular Weight: 138.17
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 194
Melting point (°C): 60


Product Categories: Bioscience, Reagents for Central Nervous System Disease Models
Other Product Names / Synonyms: Cardiazole

Formula: C₆H₁₀N₄
Boiling Pt: 194 °C
Melting Pt: 60 °C
MDL Number: MFCD00005939
CAS Number: 54-95-5
https://us.vwr.com/stibo/web/std.lang.all/07/56/28990756.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR