7-Methoxy-8-(3-methyl-2-buten-1-yl)-2H-chromen-2-one ≥98.0%
Supplier: TCI America
Synonyms:
7-Methoxy-8-(3-methylbut-2-en-1-yl)-2H-chromen-2-one
, 7-Methoxy-8-(3-methyl-2-butenyl)coumarin
, Osthole
CAS Number: 484-12-8
MDL Number: MFCD00076049
Molecular Formula: C15H16O3
Molecular Weight: 244.29
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 84
Lambda max.: 322 nm (MeOH)
Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Phenylpropanoids, Phenylpropanoids (Phytochemicals)
Other Product Names / Synonyms: 7-Methoxy-8-(3-methyl-2-butenyl)coumarin
Formula:
C15H16O3 MW: 244.2857 g/mol Melting Pt: 84 °C |
MDL Number:
MFCD00076049 CAS Number: 484-12-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...