Oxamic hydrazide ≥95.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: SemioxamazideAmino-oxamide

O0156-5G
TCO0156-5GEA 145.09 USD
TCO0156-5G
Oxamic hydrazide ≥95.0% (by titrimetric analysis)
Oxamic hydrazide

CAS Number: 515-96-8
MDL Number: MFCD00008004
Molecular Formula: C2H5N3O2
Molecular Weight: 103.08
Purity/Analysis Method: 95.0% (T)
Form: Crystal


Other Product Names / Synonyms: Amino-oxamide, Semioxamazide

Formula: C₂H₅N₃O₂
MW: 103.08 g/mol
MDL Number: MFCD00008004
CAS Number: 515-96-8
https://us.vwr.com/stibo/web/std.lang.all/04/48/28990448.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR