2,2,4,4,6,6,8,8-Octamethylcyclotetrasilazane ≥96.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 1,1,3,3,5,5,7,7-Octamethylcyclotetrasilazane

O0153-10G
TCO0153-10GEA 128.5 USD
TCO0153-10G
2,2,4,4,6,6,8,8-Octamethylcyclotetrasilazane ≥96.0% (by titrimetric analysis)
2,2,4,4,6,6,8,8-Octamethylcyclotetrasilazane

CAS Number: 1020-84-4
MDL Number: MFCD00046123
Molecular Formula: C8H28N4Si4
Molecular Weight: 292.68
Purity/Analysis Method: 96.0% (T)
Form: Crystal
Boiling point (°C): 225
Melting point (°C): 98
Flash Point (°C): 66


Other Product Names / Synonyms: Dimethylsilazane Cyclic Tetramer

Formula: C₈H₂₈N₄Si₄
MW: 292.68 g/mol
Boiling Pt: 225 °C
Melting Pt: 98 °C
Flash Pt: 66 °C
MDL Number: MFCD00046123
CAS Number: 1020-84-4
UN: 2923
ADR: 8,III
https://us.vwr.com/stibo/web/std.lang.all/04/46/28990446.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR