cis-2-Octene ≥95.0% (by GC)

Supplier: TCI America

O0098-5ML
TCO0098-5MLEA 276 USD
TCO0098-5ML
cis-2-Octene ≥95.0% (by GC)
cis-2-Octene

CAS Number: 7642-04-8
MDL Number: MFCD00065146
Molecular Formula: C8H16
Molecular Weight: 112.22
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 126
Flash Point (°C): 21
Specific Gravity (20/20): 0.73

Formula: C₈H₁₆
MW: 112.22 g/mol
Boiling Pt: 126 °C
Density: 0.73 (20 °C)
Flash Pt: 21 °C
MDL Number: MFCD00065146
CAS Number: 7642-04-8
https://us.vwr.com/stibo/web/std.lang.all/04/00/28990400.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR