Octadecyltrichlorosilane ≥85% (by GC)
Supplier: TCI America
Synonyms:
Trichloro(octadecyl)silane
O0079-25G
TCO0079-25GEA
98.25
USD
TCO0079-25G
Octadecyltrichlorosilane ≥85% (by GC)
Octadecyltrichlorosilane
CAS Number: 112-04-9
MDL Number: MFCD00000484
Molecular Formula: C18H37Cl3Si
Molecular Weight: 387.93
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 162
Flash Point (°C): 191
Freezing point (°C): 11
Specific Gravity (20/20): 0.98
Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: Octadecyltrichlorosilane, Trichloro(stearyl)silane, ODTS
Formula:
C₁₈H₃₇Cl₃Si MW: 387.94 g/mol Boiling Pt: 162 °C Density: 0.98 (20 °C) Flash Pt: 191 °C |
MDL Number:
MFCD00000484 CAS Number: 112-04-9 UN: 1800 ADR: 8,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...