3-(3-Nitrophenyl)propionic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
3-Nitrohydrocinnamic acid
N0960-25G
N0960-5G
TCN0960-25GEA
350.18
USD
TCN0960-25G
TCN0960-5G
3-(3-Nitrophenyl)propionic acid ≥98.0% (by GC, titration analysis)
3-(3-Nitrophenyl)propionic acid
CAS Number: 1664-57-9
MDL Number: MFCD01310835
Molecular Formula: C9H9NO4
Molecular Weight: 195.17
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 119
Other Product Names / Synonyms: 3-Nitrohydrocinnamic Acid
Formula:
C₉H₉NO₄ MW: 195.1721 g/mol Melting Pt: 119 °C |
MDL Number:
MFCD01310835 CAS Number: 1664-57-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...