3-Nitrochalcone ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 3-Nitrobenzylidenacetophenone

N0936-5G N0936-1G
TCN0936-5GEA 276 USD
TCN0936-5G TCN0936-1G
3-Nitrochalcone ≥98.0% (by GC)
3-Nitrochalcone

CAS Number: 614-48-2
MDL Number: MFCD00024571
Molecular Formula: C15H11NO3
Molecular Weight: 253.26
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 144


Other Product Names / Synonyms: 2-(3-Nitrobenzylidene)acetophenone

Formula: C₁₅H₁₁NO₃
MW: 253.26 g/mol
Melting Pt: 144 °C
MDL Number: MFCD00024571
CAS Number: 614-48-2
https://us.vwr.com/stibo/web/std.lang.all/01/23/28990123.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR