1,4-Naphthalenebis(trifluoromethanesulfonate) ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1,4-Naphthaleneditriflate

N0934-1G N0934-5G
TCN0934-5GEA 546 USD
TCN0934-5G TCN0934-1G
1,4-Naphthalenebis(trifluoromethanesulfonate) ≥98.0% (by GC)
1,4-Naphthalenebis(trifluoromethanesulfonate)

CAS Number: 152873-78-4
Molecular Formula: C12H6F6O6S2
Molecular Weight: 424.28
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 64
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives
Other Product Names / Synonyms: 1,4-Naphthaleneditriflate

Formula: C₁₂H₆F₆O₆S₂
MW: 424.28 g/mol
Melting Pt: 64 °C
CAS Number: 152873-78-4
https://us.vwr.com/stibo/web/std.lang.all/01/21/28990121.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR