10-(2-Naphthyl)anthracene-9-boronic acid ≥98.0% (by HPLC) (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: (10-(Naphthalen-2-yl)anthracen-9-yl)boronic acid

N0929-1G
TCN0929-1GEA 199 USD
TCN0929-1G
10-(2-Naphthyl)anthracene-9-boronic acid ≥98.0% (by HPLC) (contains varying amounts of Anhydride)
10-(2-Naphthyl)anthracene-9-boronic acid

CAS Number: 597554-03-5
MDL Number: MFCD11973625
Molecular Formula: C24H17BO2
Molecular Weight: 348.21
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal

Formula: C₂₄H₁₇BO₂
MW: 348.21 g/mol
MDL Number: MFCD11973625
CAS Number: 597554-03-5
https://us.vwr.com/stibo/web/std.lang.all/01/14/28990114.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR