Nevirapine ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 11-Cyclopropyl-5,11-dihydro-4-methyl-6H-dipyrido[3,2-b:2',3'-e][1,4]diazepin-6-one

N0922-200MG
TCN0922-200MGEA 155.3 USD
TCN0922-200MG
Nevirapine ≥98.0% (by GC)
Nevirapine

CAS Number: 129618-40-2
MDL Number: MFCD00866928
Molecular Formula: C15H14N4O
Molecular Weight: 266.30
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 246
Storage Temperature: 0°C


Product Categories: Bioscience, Antiviral Ingredients
Other Product Names / Synonyms: 11-Cyclopropyl-5,11-dihydro-4-methyl-6H-dipyrido[3,2-b:2',3'-e][1,4]diazepin-6-one

Formula: C₁₅H₁₄N₄O
MW: 266.30 g/mol
Melting Pt: 246 °C
MDL Number: MFCD00866928
CAS Number: 129618-40-2
https://us.vwr.com/stibo/web/std.lang.all/01/10/28990110.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR