Nonyl-β-D-glucopyranoside ≥97.0%
Supplier: TCI America
Synonyms:
β-Nonylglucoside
, NG
, N-Nonyl Beta-D-glucopyranoside
, (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-nonoxyoxane-3,4,5-triol
, (2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(nonyloxy)tetrahydro-2H-pyran-3,4,5-triol
CAS Number: 69984-73-2
MDL Number: MFCD00063300
Molecular Formula: C15H30O6
Molecular Weight: 306.40
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 70
Specific rotation [a]20/D: -27 deg (C=1, MeOH)
Storage Temperature:
Formula:
C₁₅H₃₀O₆ MW: 306.4 g/mol Melting Pt: 70 °C |
MDL Number:
MFCD00063300 CAS Number: 69984-73-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...