N-(1-Naphthyl)ethylenediamine dihydrochloride ≥98.0% (by HPLC, total nitrogen) for biochemical research
Supplier: TCI America
Synonyms:
N-(1-Naphthyl)ethylenediamine dihydrochloride
N0869-5G
TCN0869-5GEA
74.53
USD
TCN0869-5G
N-(1-Naphthyl)ethylenediamine dihydrochloride ≥98.0% (by HPLC, total nitrogen) for biochemical research
N-2-Aminoethyl-1-naphthylamine dihydrochloride
[for Biochemical Research]
CAS Number: 1465-25-4
MDL Number: MFCD00012556
Molecular Formula: C12H14N2
Molecular Weight: 259.17
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 190
Product Categories: Bioscience, ROS/RNS Detection Reagents, Reagents for Nitric Oxide Research
Other Product Names / Synonyms: Bratton-Marshall Reagent, Marshall's Reagent
Formula:
C₁₂H₁₆Cl₂N₂ MW: 259.18 g/mol Melting Pt: 190 °C |
MDL Number:
MFCD00012556 CAS Number: 1465-25-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...