2-(2-Naphthyl)indole ≥98.0% (by GC)
Supplier: TCI America
N0859-1G
TCN0859-1GEA
102
USD
TCN0859-1G
2-(2-Naphthyl)indole ≥98.0% (by GC)
2-(2-Naphthyl)indole
CAS Number: 23746-81-8
MDL Number: MFCD00124144
Molecular Formula: C18H13N
Molecular Weight: 243.31
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 204
Product Categories: Materials Chemistry, Ligands for Functional Metal Complexes
Other Product Names / Synonyms: 2-(2-Indolyl)naphthalene
Formula:
C₁₈H₁₃N MW: 243.31 Melting Pt: 204 °C |
MDL Number:
MFCD00124144 CAS Number: 23746-81-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...