Perfluorodecanoic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Nonadecafluorodecanoic acid

N0607-5G
TCN0607-5GEA 91.55 USD
TCN0607-5G
Perfluorodecanoic acid ≥98.0% (by titrimetric analysis)
Perfluorodecanoic acid

CAS Number: 335-76-2
MDL Number: MFCD00004175
Molecular Formula: C10HF19O2
Molecular Weight: 514.09
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Boiling point (°C): 145
Melting point (°C): 88


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks, Materials Chemistry, Fluorosurfactants
Other Product Names / Synonyms: Perfluorodecanoic Acid

Formula: C₁₀HF₁₉O₂
MW: 514.09 g/mol
Boiling Pt: 145 °C
Melting Pt: 88 °C
MDL Number: MFCD00004175
CAS Number: 335-76-2
https://us.vwr.com/stibo/web/std.lang.all/98/02/28989802.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR