(±)-δ-Nonalactone ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
(±)-Nonan-5-olide
, δ-Nonanolactone
, (±)-Tetrahydro-6-butyl-2H-pyran-2-one
, 6-Butyltetrahydro-2H-pyran-2-one
CAS Number: 3301-94-8
MDL Number: MFCD00036500
Molecular Formula: C9H16O2
Molecular Weight: 156.23
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 137
Melting point (°C): -26
Flash Point (°C): 137
Specific Gravity (20/20): 0.99
Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: delta-Nonalactone, delta-Pelargonolactone
Formula:
C₉H₁₆O₂ MW: 156.22 g/mol Boiling Pt: 137 °C Melting Pt: –26 °C Density: 0.99 (20 °C) Flash Pt: 137 °C |
MDL Number:
MFCD00036500 CAS Number: 3301-94-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...