Nonylphenol (mixture of isomers)

Supplier: TCI America

N0300-500ML N0300-25ML
TCN0300-25MLEA 25.99 USD
TCN0300-25ML TCN0300-500ML
Nonylphenol (mixture of isomers)
Nonylphenol (mixture of isomers)

CAS Number: 84852-15-3
MDL Number: MFCD00002396
Molecular Formula: C15H24O
Molecular Weight: 220.36
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 150
Specific Gravity (20/20): 0.95

Formula: C₁₅H₂₄₀
MW: 220.35 g/mol
Density: 0.95 (20 °C)
Flash Pt: 150 °C
MDL Number: MFCD00002396
CAS Number: 84852-15-3
UN: 3145
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR