Nitroguanidine ≥97.0% (by titration analysis on dried basis) wetted with ca. 25% water
Supplier: TCI America
Synonyms:
N-Nitroguanidine
N0206-25G
TCN0206-25GEA
28.4
USD
TCN0206-25G
Nitroguanidine ≥97.0% (by titration analysis on dried basis) wetted with ca. 25% water
Nitroguanidine
CAS Number: 556-88-7
MDL Number: MFCD00007039
Molecular Formula: CH4N4O2
Molecular Weight: 104.07
Purity/Analysis Method:
Form: Crystal
Formula:
CH₄N₄O₂ |
MDL Number:
MFCD00007039 CAS Number: 556-88-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...