1,5-Diisocyanatonaphthalene ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
Naphthalene 1,5-Diisocyanate
N0168-25G
TCN0168-25GEA
151.19
USD
TCN0168-25G
1,5-Diisocyanatonaphthalene ≥98.0% (by GC)
1,5-Diisocyanatonaphthalene
CAS Number: 3173-72-6
Molecular Formula: C12H6N2O2
Molecular Weight: 210.19
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 183
Melting point (°C): 129
Flash Point (°C): 192
Storage Temperature: 0-10°C
Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: Naphthalene 1,5-Diisocyanate
Formula:
C₁₂H₆N₂O₂ MW: 210.19 g/mol Boiling Pt: 183 °C Melting Pt: 129 °C Flash Pt: 192 °C |
CAS Number:
3173-72-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...