Norbornene ≥99.0% (by GC)
Supplier: TCI America
Synonyms:
Norbornylene
N0166-400G
N0166-25G
TCN0166-400GEA
90.83
USD
TCN0166-400G
TCN0166-25G
Norbornene ≥99.0% (by GC)
Norbornene
CAS Number: 498-66-8
MDL Number: MFCD00082304
Molecular Formula: C7H10
Molecular Weight: 94.16
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 96
Melting point (°C): 47
Flash Point (°C): -14
Product Categories: Synthetic Organic Chemistry, C-H Bond Activation Reaction, Ligands
Other Product Names / Synonyms: Bicyclo[2.2.1]hept-2-ene
Formula:
C₇H₁₀ Boiling Pt: 96 °C Melting Pt: 47 °C Flash Pt: –14 °C |
MDL Number:
MFCD00082304 CAS Number: 498-66-8 UN: 1325 ADR: 4.1,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...