2-Methylisonicotinic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-Methylpyridine-4-carboxylic acid

M2602-1G
TCM2602-1GEA 183 USD
TCM2602-1G
2-Methylisonicotinic acid ≥98.0% (by GC, titration analysis)
2-Methylisonicotinic acid

CAS Number: 4021-11-8
MDL Number: MFCD00513435
Molecular Formula: C7H7NO2
Molecular Weight: 137.14
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 294


Other Product Names / Synonyms: 2-Methyl-4-pyridinecarboxylic Acid

Formula: C₇H₇NO₂
MW: 137.14
Melting Pt: 294 °C
MDL Number: MFCD00513435
CAS Number: 4021-11-8
https://us.vwr.com/stibo/web/std.lang.all/86/26/28988626.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR