Methyl-3-(2-bromophenyl)propionate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Methyl 3-(2-bromophenyl)propanoate3-(2-Bromophenyl)propionic Acid Methyl Ester

M2587-1G M2587-5G
TCM2587-1GEA 30.45 USD
TCM2587-1G TCM2587-5G
Methyl-3-(2-bromophenyl)propionate ≥98.0% (by GC)
Methyl-3-(2-bromophenyl)propionate

CAS Number: 66191-86-4
MDL Number: MFCD09953149
Molecular Formula: C10H11BrO2
Molecular Weight: 243.10
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 87
Specific Gravity (20/20): 1.41
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 3-(2-Bromophenyl)propionic Acid Methyl Ester

Formula: C₁₀H₁₁BrO₂
MW: 243.10 g/mol
Boiling Pt: 87 °C
Density: 1.41 (20 °C)
MDL Number: MFCD09953149
CAS Number: 66191-86-4
https://us.vwr.com/stibo/web/std.lang.all/86/14/28988614.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR