Metoprolol tartrate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 1-(Isopropylamino)-3-[4-(beta-methoxyethyl)phenoxy]-2-propanol (+)-Tartrate

M2555-25G M2555-5G
TCM2555-25GEA 259 USD
TCM2555-25G TCM2555-5G
Metoprolol tartrate ≥98.0% (by HPLC, titration analysis)
Metoprolol tartrate

CAS Number: 56392-17-7
MDL Number: MFCD00056257
Molecular Formula: C15H25NO3
Molecular Weight: 342.41
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 123
Specific rotation [a]20/D: 9 deg (C=2, H2O)
Lambda max.: 223 nm (H2O)


Product Categories: Bioscience, Antihypertensive Ingredients
Other Product Names / Synonyms: 1-(Isopropylamino)-3-[4-(beta-methoxyethyl)phenoxy]-2-propanol (+)-Tartrate

Formula: C₁₅H₂₅NO₃
MW: 342.41 g/mol
Melting Pt: 123 °C
MDL Number: MFCD00056257
CAS Number: 56392-17-7
https://us.vwr.com/stibo/web/std.lang.all/85/86/28988586.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR