2-Morpholino-4H-pyrimido[2,1-a]isoquinolin-4-one ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Compound 401

M2537-10MG M2537-50MG
TCM2537-10MGEA 92 USD
TCM2537-10MG TCM2537-50MG
2-Morpholino-4H-pyrimido[2,1-a]isoquinolin-4-one ≥98.0% (by GC)
2-Morpholino-4H-pyrimido[2,1-a]isoquinolin-4-one

CAS Number: 168425-64-7
MDL Number: MFCD10568147
Molecular Formula: C16H15N3O2
Molecular Weight: 281.32
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 214
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Compound 401

Formula: C₁₆H₁₅N₃O₂
MW: 281.32 g/mol
Melting Pt: 214 °C
MDL Number: MFCD10568147
CAS Number: 168425-64-7
https://us.vwr.com/stibo/web/std.lang.all/85/67/28988567.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR