2-Methoxyestradiol ≥97.0% (by GC)

Supplier: TCI America

M2530-100MG M2530-25MG
TCM2530-25MGEA 44.6 USD
TCM2530-25MG TCM2530-100MG
2-Methoxyestradiol ≥97.0% (by GC)
2-Methoxyestradiol

CAS Number: 362-07-2
MDL Number: MFCD00010489
Molecular Formula: C19H26O3
Molecular Weight: 302.41
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Melting point (°C): 187
Specific rotation [a]20/D: 107 deg (C=0.5, THF)
Lambda max.: 286 nm (EtOH)

Formula: C₁₉H₂₆O₃
Melting Pt: 187 °C
MDL Number: MFCD00010489
CAS Number: 362-07-2
https://us.vwr.com/stibo/web/std.lang.all/85/62/28988562.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR